Learn More
Grainger Fiberglass Stair Tread 48 x 12 In
Catalog No. 502766002
Encompass_Preferred
The Fibergrate Fiberglass Bar Stair Tread is a heavy duty stair tread made of molded fiberglass reinforced plastic with a meniscus bar surface. It has a slip resistant surface and is designed for industrial and commercial use on stairs and landings. This stair tread measures 36 inches long x 11-1/2 inches wide x 1-3/8 inches thick and weighs approximately 8 lbs. It can support a uniform load of 100 psf. The bar spacing is 1-3/16 inches on center. This stair tread meets OSHA standards for slip resistance on stairs. It comes in safety yellow color.
Catalog No. 50-276-6002 Supplier Grainger Supplier No. 16U526
The Fibergrate Fiberglass Bar Stair Tread is a heavy duty stair tread made of molded fiberglass reinforced plastic with a meniscus bar surface. It has a slip resistant surface and is designed for industrial and commercial use on stairs and landings. This stair tread measures 36 inches long x 11-1/2 inches wide x 1-3/8 inches thick and weighs approximately 8 lbs. It can support a uniform load of 100 psf. The bar spacing is 1-3/16 inches on center. This stair tread meets OSHA standards for slip resistance on stairs. It comes in safety yellow color.