Learn More
MSC PrecisionAire 10055.011625 16 x 25 x 1", MERV 4, <20% Efficiency, Panel Air Filter Fiberglass, Chipboard Frame, 300 Max FPM, 833 CFM, Use with Any Unit
Supplier:  MSC 98674484
Encompass
PrecisionAire 10055.011625 16 x 25 x 1", MERV 4, <20% Efficiency, Panel Air Filter Fiberglass, Chipboard Frame, 300 Max FPM, 833 CFM, Use with Any Unit
Catalog No. 50-192-0208