Learn More
Thermo Scientific Fiberlite 500mL Bottle 2/pack
Supplier:  Thermo Scientific 0101493
Encompass_Preferred
Thermo Scientific™ 500mL Fiberlite Bottles combine a Nalgene bottle body with newly designed Fiberlite Cap Closure. These bottles will be available for Thermo Scientific™ Sorvall™ LYNX Superspeed
Catalog No. NC1455535