Learn More
Thermo Scientific™ Fiberlite™ F14-6 x 250y Adapters
Catalog No. 0101117 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity:
10 mL Vacutainer Tube
100 mL Oak Ridge Tube
15 mL Conical Tube
16 mL Oak Ridge Tube
3 mL Vacutainer Tube
30 mL Oak Ridge Tube
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
01-011-17 10 mL Vacutainer Tube
01-010-72 30 mL Oak Ridge Tube
01-010-74 16 mL Oak Ridge Tube
01-011-19 100 mL Oak Ridge Tube
01-011-38 3 mL Vacutainer Tube
01-014-10 15 mL Conical Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 01-011-17 Supplier Thermo Scientific™ Supplier No. 0101117
Add to Cart
Add to Cart
Add versatility to your fixed-angle rotor with Thermo Scientific™ Fiberlite™ F14-6 x 250y Adapters, which support various conical tubes.
Add versatility to your fixed-angle rotor with Thermo Scientific™ Fiberlite™ F14-6 x 250y Adapters, which support Ridge™, Vacutainer™ and conical tubes.
Adapters hold Oak Ridge™, Vacutainer™ and conical tubes, and vary by capacity.
Ordering Information:
Order Lid O-ring (022062034), O-ring refresher kit (021062038), Lid assembly (099062034) and Fiberlite F14-6x250y Rotor (78500) separately. Contact your Sales Representative for more information.
TRUSTED_SUSTAINABILITY
Specifications
Type Rotor Tube Adapter
Capacity 10 mL Vacutainer Tube
Vessels per Adapter 7