Learn More
Thermo Scientific™ Fiberlite™ H3-LV Large Volume Swinging Bucket Rotor
GREENER_CHOICE
Catalog No. 75003665 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Thermo Scientific Sorvall Legend XT/XF, Multifuge X3/X3F and SL 40F Centrifuges
Catalog No. For Use With (Equipment)
75-003-665 Thermo Scientific Sorvall Legend XT/XF, Multifuge X3/X3F and SL 40F Centrifuges
1 options
Catalog No. 75-003-665 Supplier Thermo Scientific™ Supplier No. 75003665
Add to Cart
Add to Cart
Request a Quote
Conduct high-volume microplate processing with this large volume swinging bucket rotor.
Conduct high-volume microplate processing with the Thermo Scientific™ Fiberlite™ H3-LV Large Volume Swinging Bucket Rotor.
Features:
Fiberlite rotors:
  • Lightweight carbon fiber rotors reduce run-times by achieving higher G-forces
  • Corrosion-and fatigue-resistant design ensures structural integrity
  • Rotors are backed by a 15-year warranty and are compatible with Thermo Scientific™ Nalgene™ bottles and tubes for complete centrifuge solutions
Recommended for:
Pelleting cells and cellular debris, protein precipitation, plasmid purification and collecting physiological fluids for diagnostic tests
Note:
†Performance may vary by centrifuge model.
TRUSTED_SUSTAINABILITY
Specifications
Description Fiberlite H3-LV Large Volume Swinging Bucket Rotor for Thermo Scientific Sorvall Legend XT, Multifuge X3 Series
Type Swinging Bucket Rotor
Capacity 28 Standard or 8 Deepwell Plates
Max. RCF 1,840 x g
Max. Speed 3,600 rpm
For Use With (Equipment) Thermo Scientific Sorvall Legend XT/XF, Multifuge X3/X3F and SL 40F Centrifuges
Angle 90°
Biocontainment Yes