Learn More
Medchemexpress LLC HY-15874A 5mg Medchemexpress, Fiboflapon sodium CAS:1196070-26-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY15874A5MG
Encompass_Preferred
Medchemexpress, HY-15874A 5mg Fiboflapon sodium CAS:1196070-26-4 Fiboflapon sodium (GSK2190915; AM-803) is a potent and orally bioavailable 5-lipoxygenase-activating protein (FLAP) inhibitor with a potency of 2.9 nM in FLAP binding, an IC50 of 76 nM for inhibition of LTB4 in human blood. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1812