Learn More
MSC Fibre-Metal by PIP Hard Hat, Class G, 8-Point Suspension, White, Fiberglass
Catalog No. 5000509402
Encompass
This standard hard hat is designed for general use, featuring a Class G rating and an 8-point suspension system for comfort and protection. Constructed from durable fiberglass, it comes in a white finish with a ratchet adjustment for a secure fit.
  • Class G rated for electrical protection.
  • 8-point suspension system enhances comfort and impact absorption.
  • Fiberglass material provides durability.
  • Ratchet adjustment ensures a secure and customizable fit.
  • ANSI Type I compliance for top impact protection.
  • Quick reversing suspension for versatility.
  • Non-vented design for industrial environments.
  • Universal head size fit.
Catalog No. 50-005-09402 Supplier MSC Supplier No. 44508174
This standard hard hat is designed for general use, featuring a Class G rating and an 8-point suspension system for comfort and protection. Constructed from durable fiberglass, it comes in a white finish with a ratchet adjustment for a secure fit.
  • Class G rated for electrical protection.
  • 8-point suspension system enhances comfort and impact absorption.
  • Fiberglass material provides durability.
  • Ratchet adjustment ensures a secure and customizable fit.
  • ANSI Type I compliance for top impact protection.
  • Quick reversing suspension for versatility.
  • Non-vented design for industrial environments.
  • Universal head size fit.