Learn More
MSC Fibre-Metal Faceshield Window, Noryl, 7 in Crown, for Protective Cap
Catalog No. 5000507791
Encompass
The Fibre-Metal F5500BP High Performance Faceshield Window is constructed from Noryl, designed for attachment to a protective cap. It features a 7-inch crown and a mounting loop, and is supplied in bulk packaging with 25 units per box.
  • Constructed from Noryl material.
  • Designed for use with a protective cap.
  • Includes a 7 in crown.
  • Features a mounting loop.
  • Packaged in bulk with 25 units per box.
  • High performance design.
Catalog No. 50-005-07791 Supplier MSC Supplier No. 82609371
The Fibre-Metal F5500BP High Performance Faceshield Window is constructed from Noryl, designed for attachment to a protective cap. It features a 7-inch crown and a mounting loop, and is supplied in bulk packaging with 25 units per box.
  • Constructed from Noryl material.
  • Designed for use with a protective cap.
  • Includes a 7 in crown.
  • Features a mounting loop.
  • Packaged in bulk with 25 units per box.
  • High performance design.