Learn More
MSC Fibre-Metal Hard Hat, Class E, 8-Point Suspension, Gray, Plastic
Catalog No. 5000508491
Encompass
This hard hat is designed for head protection, featuring a Class E rating and an 8-point suspension system. It comes in a gray color and is constructed from durable plastic.
  • Standard type
  • Gray color
  • Ratchet adjustment type
  • ANSI Type I
  • Plastic material
  • Not vented
  • No hard hat graphics
  • Class E rating
  • Quick reversing suspension
  • Nylon suspension material
  • Not slotted
  • Universal head size fit
  • 8-point suspension
Catalog No. 50-005-08491 Supplier MSC Supplier No. 64302763
This hard hat is designed for head protection, featuring a Class E rating and an 8-point suspension system. It comes in a gray color and is constructed from durable plastic.
  • Standard type
  • Gray color
  • Ratchet adjustment type
  • ANSI Type I
  • Plastic material
  • Not vented
  • No hard hat graphics
  • Class E rating
  • Quick reversing suspension
  • Nylon suspension material
  • Not slotted
  • Universal head size fit
  • 8-point suspension