Learn More
MSC Fibre-Metal Hard Hat, Class E, 8-Point Suspension, White, Ratchet Adjustment
Catalog No. 5000508259
Encompass
This hard hat is designed for standard industrial applications. It features a plastic construction, a white finish, and an 8-point nylon suspension system for comfort and protection. The ratchet adjustment type ensures a secure fit for head sizes from 6-1/2 to 8, meeting Class E and ANSI Type I safety standards.
  • Standard type hard hat
  • White color
  • Ratchet adjustment for a secure fit
  • Made from durable plastic
  • Meets ANSI type I standards
  • Class E rated for electrical hazards
  • Not vented or slotted
  • 8-point nylon suspension for impact protection
  • Accommodates head sizes 6-1/2 to 8
Catalog No. 50-005-08259 Supplier MSC Supplier No. 64302540
This hard hat is designed for standard industrial applications. It features a plastic construction, a white finish, and an 8-point nylon suspension system for comfort and protection. The ratchet adjustment type ensures a secure fit for head sizes from 6-1/2 to 8, meeting Class E and ANSI Type I safety standards.
  • Standard type hard hat
  • White color
  • Ratchet adjustment for a secure fit
  • Made from durable plastic
  • Meets ANSI type I standards
  • Class E rated for electrical hazards
  • Not vented or slotted
  • 8-point nylon suspension for impact protection
  • Accommodates head sizes 6-1/2 to 8