Learn More
MSC Fibre-Metal Hard Hat, Class E, Full Brim, Hi-Viz Orange, Ratchet Suspension
Catalog No. 5000507984
Encompass
This hard hat is designed to provide head protection in various industrial environments. It features a full brim and a ratchet adjustment system for a secure and comfortable fit.
  • Class E rating for electrical protection
  • Full brim type for added coverage
  • Hi-Viz orange color for enhanced visibility
  • Ratchet adjustment for a secure fit
  • Made from durable plastic material
  • 8-point suspension system
  • Universal fit for various head sizes
Catalog No. 50-005-07984 Supplier MSC Supplier No. 64302326
This hard hat is designed to provide head protection in various industrial environments. It features a full brim and a ratchet adjustment system for a secure and comfortable fit.
  • Class E rating for electrical protection
  • Full brim type for added coverage
  • Hi-Viz orange color for enhanced visibility
  • Ratchet adjustment for a secure fit
  • Made from durable plastic material
  • 8-point suspension system
  • Universal fit for various head sizes