Learn More
MSC Fibre-Metal Hard Hat, Full Brim, Class E, 8-Point Suspension, Ratchet, White
Catalog No. 5000508475
Encompass
Fibre-Metal hard hats are designed for worker protection and comfort. This full brim hard hat features a Class E rating for electrical protection and an 8-point suspension system with a ratchet adjustment for a secure and comfortable fit.
  • Full brim design provides additional protection.
  • Class E rating ensures electrical protection.
  • 8-point suspension system for comfort and safety.
  • Ratchet adjustment allows for a secure and customized fit.
  • Constructed from durable plastic.
  • Universal fit accommodates various head sizes.
  • Non-vented for maximum protection.
Catalog No. 50-005-08475 Supplier MSC Supplier No. 64302276
Fibre-Metal hard hats are designed for worker protection and comfort. This full brim hard hat features a Class E rating for electrical protection and an 8-point suspension system with a ratchet adjustment for a secure and comfortable fit.
  • Full brim design provides additional protection.
  • Class E rating ensures electrical protection.
  • 8-point suspension system for comfort and safety.
  • Ratchet adjustment allows for a secure and customized fit.
  • Constructed from durable plastic.
  • Universal fit accommodates various head sizes.
  • Non-vented for maximum protection.