Learn More
MSC Fibre-Metal F500 Face Shield Crown, Gray, 7 in. Crown, 1 ea.
Catalog No. 5000509251
Encompass
This gray high-performance headgear is designed to provide crown protection for faceshield visors. It features a lightweight, durable construction that is more heat resistant than ordinary alternatives, while offering integrated space for air circulation to enhance wearer comfort.
  • Features 7 in. crown protection
  • Equipped with a ratchet suspension for adjustable fit
  • Accommodates standard size windows
  • Meets ANSI Z87.1 and CSA Z94.3 standards
  • Compatible with a wide range of faceshield visors
Catalog No. 50-005-09251 Supplier MSC Supplier No. 94303336
This gray high-performance headgear is designed to provide crown protection for faceshield visors. It features a lightweight, durable construction that is more heat resistant than ordinary alternatives, while offering integrated space for air circulation to enhance wearer comfort.
  • Features 7 in. crown protection
  • Equipped with a ratchet suspension for adjustable fit
  • Accommodates standard size windows
  • Meets ANSI Z87.1 and CSA Z94.3 standards
  • Compatible with a wide range of faceshield visors