Learn More
Honeywell™ Fibre-Metal™ Homerun™ Bump Caps
Catalog No. 191600077
Change view
Click to view available options
Bump cap; Black
Catalog No.
19-160-0077 Bump cap; Black
1 options
This item is not returnable. View return policy
Catalog No. 19-160-0077 Supplier Honeywell™ Supplier No. SBC2BK
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Style and comfort of a quality baseball cap with the protection of a thermoplastic inner liner to create a bump cap that protects from minor lacerations, abrasions, and bruises.
  • Cooling ventilation eyelets permit air circulation.
  • Adjustable back strap and inner surface foam pad for extra comfort.
  • Not ANSI/CSA approved.
TRUSTED_SUSTAINABILITY
Specifications
Color Black