Learn More
MSC Fibre-Metal Replacement Window, Green, 0.06 inch, 19 inch
Catalog No. 5000507798
Encompass
This product is a replacement window designed for use with specific series face shields and aluminum cap brackets. It meets CSA Z94.3 and ANSI Z87.1+ standards.
  • Replacement window
  • Bright/uncoated
  • No lens shade
  • 19 inch window width
  • 0.06 inch window thickness
  • Compatible with F400 series, F500 series, and FH66 aluminum cap brackets
  • Meets ANSI Z87.1+ standard
  • Meets CSA Z94.3 standard
Catalog No. 50-005-07798 Supplier MSC Supplier No. 92144013
This product is a replacement window designed for use with specific series face shields and aluminum cap brackets. It meets CSA Z94.3 and ANSI Z87.1+ standards.
  • Replacement window
  • Bright/uncoated
  • No lens shade
  • 19 inch window width
  • 0.06 inch window thickness
  • Compatible with F400 series, F500 series, and FH66 aluminum cap brackets
  • Meets ANSI Z87.1+ standard
  • Meets CSA Z94.3 standard