Learn More
Advanced Biomatrix FibriCol, Bovine Collagen Solution, 10 mg/mL, 20 mL, Atelocollagen
Catalog No. 501960278
Encompass_Preferred
FibriCol collagen is known as the standard of all collagens for purity (>99% collagen content), functionality, and the most native-like collagen available. FibriCol is isolated from bovine hides sourced from the only controlled, closed herd in the United States. Advanced BioMatrix's manufacturing processes comply with stringent quality standards that have proven to yield unsurpassed lot-to-lot consistency. FibriCol collagen is approximately 97% Type I collagen with the remainder being comprised of Type III collagen. It contains a high monomer content as measured by gel permeation chromatography. FibriCol collagen is supplied at approximately a 10 mg/mL concentration. FibriCol is soluble atelo-collagen in 0.01 N HCI, therefore, the pH is approximately 2.0. FibriCol collagen is ideal for applications where high concentrations of collagen are needed. FibriCol is sterile filtered and is supplied as a ready to use solution. Collagen Training Video: https://youtu.be/YcK2vUpNt0g
Catalog No. 50-196-0278 Supplier Advanced Biomatrix Supplier No. 513320ML
FibriCol collagen is known as the standard of all collagens for purity (>99% collagen content), functionality, and the most native-like collagen available. FibriCol is isolated from bovine hides sourced from the only controlled, closed herd in the United States. Advanced BioMatrix's manufacturing processes comply with stringent quality standards that have proven to yield unsurpassed lot-to-lot consistency. FibriCol collagen is approximately 97% Type I collagen with the remainder being comprised of Type III collagen. It contains a high monomer content as measured by gel permeation chromatography. FibriCol collagen is supplied at approximately a 10 mg/mL concentration. FibriCol is soluble atelo-collagen in 0.01 N HCI, therefore, the pH is approximately 2.0. FibriCol collagen is ideal for applications where high concentrations of collagen are needed. FibriCol is sterile filtered and is supplied as a ready to use solution. Collagen Training Video: https://youtu.be/YcK2vUpNt0g