Learn More
Sigma Aldrich Fine Chemicals Biosciences fibriColR protein from bov
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 513320ML
Encompass
fibriCol(R) collagen is approximately 97% Type I collagen with the remainder being comprised of Type III collagen. It contains a high monomer content as measured by gel permeation chromatography.It is ideal for applications where high concentrations of collagen are needed. fibriCol(R) collagen is provided in a 20 mL volume and is contained in user-friendly packaging for use and storage. fibriCol(R) is sterile filtered and is supplied as a ready to use SOLUTIONon.
Catalog No. 50-290-1058