Learn More
ABclonal Technology Fibrillarin Rabbit mAb
SDP
Catalog No. 5000014157
Encompass_Preferred
This gene product is a component of a nucleolar small nuclear ribonucleoprotein (snRNP) particle thought to participate in the first step in processing preribosomal RNA It is associated with the U3 U8 and U13 small nuclear RNAs and is located in the dense fibrillar component (DFC) of the nucleolus The encoded protein contains an N-terminal repetitive domain that is rich in glycine and arginine residues like fibrillarins in other species Its central region resembles an RNA-binding domain and contains an RNP consensus sequence Antisera from approximately 8% of humans with the autoimmune disease scleroderma recognize fibrillarin
Catalog No. 50-000-14157 Supplier ABclonal Technology Supplier No. A2781820UL
This gene product is a component of a nucleolar small nuclear ribonucleoprotein (snRNP) particle thought to participate in the first step in processing preribosomal RNA It is associated with the U3 U8 and U13 small nuclear RNAs and is located in the dense fibrillar component (DFC) of the nucleolus The encoded protein contains an N-terminal repetitive domain that is rich in glycine and arginine residues like fibrillarins in other species Its central region resembles an RNA-binding domain and contains an RNP consensus sequence Antisera from approximately 8% of humans with the autoimmune disease scleroderma recognize fibrillarin