Learn More
Medchemexpress LLC HY-P1507 1mg Medchemexpress, Fibrinogen Binding Inhibitor Peptide CAS:89105-94-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP15071MG
Encompass_Preferred
Medchemexpress, HY-P1507 1mg Fibrinogen Binding Inhibitor Peptide CAS:89105-94-2 Fibrinogen Binding Inhibitor Peptide is a dodecapeptide (HHLGGAKQAGDV, H12), which is a fibrinogen γ-chain carboxy-terminal sequence (γ400-411). Fibrinogen Binding Inhibitor Peptide is a specific binding site of the ligand for activated glycoprotein (GP) IIb/IIIa. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3062