Learn More
Medchemexpress LLC Fibrinogen-binding peptide | 137235-80-4 | 96.32% | 565.62 | 10 MG
SDP
Catalog No. 5000423167
Encompass_Preferred
Fibrinogen-Binding Peptide is a presumptive peptide mimic of the vitronectin binding site on the fibrinogen receptor. It binds fibrinogen and inhibits both the adhesion of platelets to fibrinogen and platelet aggregation, and also inhibits the adhesion of platelets to vitronectin.
  • Shipping at room temperature in continental US; may vary elsewhere.
  • For solid storage, keep sealed, away from moisture.
  • Solid storage stability: 2 years at -80°C, 1 year at -20°C.
  • For in-solvent storage, keep sealed, away from moisture.
  • In-solvent storage stability: 6 months at -80°C, 1 month at -20°C.
Catalog No. 50-004-23167 Supplier Medchemexpress LLC Supplier No. HYP174110MG
Fibrinogen-Binding Peptide is a presumptive peptide mimic of the vitronectin binding site on the fibrinogen receptor. It binds fibrinogen and inhibits both the adhesion of platelets to fibrinogen and platelet aggregation, and also inhibits the adhesion of platelets to vitronectin.
  • Shipping at room temperature in continental US; may vary elsewhere.
  • For solid storage, keep sealed, away from moisture.
  • Solid storage stability: 2 years at -80°C, 1 year at -20°C.
  • For in-solvent storage, keep sealed, away from moisture.
  • In-solvent storage stability: 6 months at -80°C, 1 month at -20°C.