Learn More
Medchemexpress LLC Fibrinogen-Binding Peptide | 137235-80-4 | 99.8% | 565.62 | 5 MG
SDP
Catalog No. 5000423621
Encompass_Preferred
Fibrinogen-Binding Peptide is a presumptive peptide mimic of the vitronectin binding site on the fibrinogen receptor, designed by the anticomplementarity hypothesis. It binds to fibrinogen and acts to inhibit both the adhesion of platelets to fibrinogen and platelet aggregation. Additionally, it inhibits the adhesion of platelets to vitronectin.
  • Inhibits platelet adhesion to fibrinogen
  • Prevents platelet aggregation
  • Inhibits platelet adhesion to vitronectin
Catalog No. 50-004-23621 Supplier Medchemexpress LLC Supplier No. HYP17415MG
Fibrinogen-Binding Peptide is a presumptive peptide mimic of the vitronectin binding site on the fibrinogen receptor, designed by the anticomplementarity hypothesis. It binds to fibrinogen and acts to inhibit both the adhesion of platelets to fibrinogen and platelet aggregation. Additionally, it inhibits the adhesion of platelets to vitronectin.
  • Inhibits platelet adhesion to fibrinogen
  • Prevents platelet aggregation
  • Inhibits platelet adhesion to vitronectin