Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Fibrinogen, Bovine

Catalog No. p-5386214
Change view
Click to view available options
Quantity:
1g
5g
10g
25g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6327603 1g
AAJ6327606 5g
AAJ6327609 10g
AAJ6327614 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAJ6327603 Supplier Thermo Scientific Chemicals Supplier No. J6327603
Only null left
Add to Cart

Fibrinogen (CAS # 9001-32-5) is a glycoprotein produced in the vertebrate liver that aids in clot formation.

A soluble plasma glycoprotein, produced by the liver, that is converted by thrombin into fibrin during the blood coagulation process. Fibrinogen is a natural blood protein involved in the coagulation process. It is used instead of traditional sources of fibrinogen such as the blood products, fresh frozen plasma and cryoprecipitate. It helps in blood clot formation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Fibrinogen exists in blood as a dimer of three bonded, non-identical chains, FGA, FGG, and FGB

• Fibrinogen is cleaved by thrombin to form fibrin, the most abundant component of blood clots

Application

• Fibrinogen can be used in a laboratory setting to form polymer hydrogels, particularly for use as a tissue scaffold

• Fibrinogen may be useful in the research of neurodegeneration and brain injury; some evidence shows that it has a signaling role in neural progenitor cells

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Neural Signaling
Formulation Lyophilized powder
Quantity 1g
Source Bovine Serum
Storage Requirements 2° to 8°C
Regulatory Status RUO
Product Type Fibrinogen, Bovine
Recombinant Native
Form Lyophilized

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.