Learn More
Medchemexpress LLC Fibrinogen (bovine) | 9001-32-5 | 92.0% | ≈340 kDa | 100MG
SDP
Catalog No. 5000238343
Encompass_Preferred
Bovine fibrinogen is a plasma glycoprotein supplied as a white to off-white powder for research applications in coagulation, thrombosis, and vascular biology. It is converted to fibrin by thrombin and is used in in vitro clotting assays and studies of inflammatory signaling in endothelial cells.
  • White to off-white powder; soluble plasma glycoprotein.
  • Used to study blood clotting, fibrin formation, thrombosis, and vascular inflammation.
  • Activated by thrombin to assemble fibrin clots.
  • Suitable for in vitro clotting assays and endothelial cell signaling studies.
  • Stored per certificate of analysis; typically kept frozen for long-term storage (e.g., -20°C).
  • Documentation available: datasheet, certificate of analysis, safety data sheet, and handling instructions.
  • CAS number: 9001-32-5.
Catalog No. 50-002-38343 Supplier Medchemexpress LLC Supplier No. HY125864100MG
Bovine fibrinogen is a plasma glycoprotein supplied as a white to off-white powder for research applications in coagulation, thrombosis, and vascular biology. It is converted to fibrin by thrombin and is used in in vitro clotting assays and studies of inflammatory signaling in endothelial cells.
  • White to off-white powder; soluble plasma glycoprotein.
  • Used to study blood clotting, fibrin formation, thrombosis, and vascular inflammation.
  • Activated by thrombin to assemble fibrin clots.
  • Suitable for in vitro clotting assays and endothelial cell signaling studies.
  • Stored per certificate of analysis; typically kept frozen for long-term storage (e.g., -20°C).
  • Documentation available: datasheet, certificate of analysis, safety data sheet, and handling instructions.
  • CAS number: 9001-32-5.