Learn More
Medchemexpress LLC Fibrinogen (bovine) | 9001-32-5 | 340 kDa | 250MG
SDP
Catalog No. 5000236115
Encompass_Preferred
Fibrinogen (Bovine) is a purified bovine plasma fibrinogen for research use, suitable for studies of fibrin assembly, blood clotting, thrombosis, and vascular inflammation. It can be activated by thrombin to form fibrin clots and has been reported to regulate NF-κB signaling and chemokine expression in endothelial cells. Supplied as a white to off-white solid; consult the Certificate of Analysis for lot-specific purity and storage conditions.
  • Purified bovine fibrinogen for in vitro research.
  • Supports fibrin clot formation when activated by thrombin.
  • Useful for studies of blood clotting, thrombosis, and vascular disease.
  • Modulates NF-κB activation and inflammatory chemokine expression.
  • Supplied as a white to off-white solid powder.
  • Consult certificate of analysis for purity and storage information.
Catalog No. 50-002-36115 Supplier Medchemexpress LLC Supplier No. HY125864250MG
Fibrinogen (Bovine) is a purified bovine plasma fibrinogen for research use, suitable for studies of fibrin assembly, blood clotting, thrombosis, and vascular inflammation. It can be activated by thrombin to form fibrin clots and has been reported to regulate NF-κB signaling and chemokine expression in endothelial cells. Supplied as a white to off-white solid; consult the Certificate of Analysis for lot-specific purity and storage conditions.
  • Purified bovine fibrinogen for in vitro research.
  • Supports fibrin clot formation when activated by thrombin.
  • Useful for studies of blood clotting, thrombosis, and vascular disease.
  • Modulates NF-κB activation and inflammatory chemokine expression.
  • Supplied as a white to off-white solid powder.
  • Consult certificate of analysis for purity and storage information.