Learn More
Siemens Healthineers Fibrinogen Calibrator Kit (Levels 1 to 6)
Catalog No. 23044680 Shop All Siemens Healthineers Products
Change view
Click to view available options
Quantity:
6 x 1 mL (1 for each level)
Catalog No. Quantity
23-044-680 6 x 1 mL (1 for each level)
1 options
This item is not returnable. View return policy
Catalog No. 23-044-680 Supplier Siemens Healthineers Supplier No. 10446148
Add to Cart
Add to Cart
This item is not returnable. View return policy
Calibrators for the assay of fibrinogen by the method of Clauss using Multifibren U.
The Fibrinogen Calibrator Kit comprises a set of six plasmas. Fibrinogen Calibrators 1 to 6 are used to prepare reference curves for the fibrinogen assay by the Clauss method using the our Multifibren U Reagent. (Fibrinogen levels 1 to 6 have a range of approximately 0.6 to 9.0 g/L.)
TRUSTED_SUSTAINABILITY
Specifications
Type Coagulation Control
For Use With (Application) Fibrinogen Testing
Level 1 to 6
Quantity 6 x 1 mL (1 for each level)