Learn More
Cusabio Technology LLC Fibrinogen Degradation Product
Catalog No. 5000050787
Encompass_Preferred
This kit detects Fibrinogen Degradation Product (FDP) in rat serum plasma and tissue homogenates using a sandwich ELISA method It provides a quantitative detection range of 15 6 ng/mL to 1000 ng/mL with a sensitivity of 3 9 ng/mL ensuring accurate measurement of low-concentration samples The assay requires a sample volume of 50-100 L and can be completed within 1-5 hours Results are measured at an optical density of 450 nm offering a reliable and efficient solution for assessing fibrinolytic activity in experimental models The kit is optimized for both precision and practicality making it suitable for research applications requiring robust quantification of FDP levels
Catalog No. 50-000-50787 Supplier Cusabio Technology LLC Supplier No. CSBE07942R
This kit detects Fibrinogen Degradation Product (FDP) in rat serum plasma and tissue homogenates using a sandwich ELISA method It provides a quantitative detection range of 15 6 ng/mL to 1000 ng/mL with a sensitivity of 3 9 ng/mL ensuring accurate measurement of low-concentration samples The assay requires a sample volume of 50-100 L and can be completed within 1-5 hours Results are measured at an optical density of 450 nm offering a reliable and efficient solution for assessing fibrinolytic activity in experimental models The kit is optimized for both precision and practicality making it suitable for research applications requiring robust quantification of FDP levels