Learn More
Cusabio Technology LLC Fibrinogen Degradation Product
Catalog No. 5000050536
Encompass_Preferred
This kit detects Fibrinogen Degradation Product (FDP) in human serum cell culture supernates urine and tissue homogenates using a quantitative sandwich ELISA method Designed for high sensitivity it offers a detection range of 6 25 ng/mL to 400 ng/mL with a lower limit of 1 56 ng/mL ensuring accurate measurement of low-abundance samples The assay requires a sample volume of 50-100 L and can be completed within 1-5 hours Absorbance is measured at 450 nm for result interpretation making it suitable for research and diagnostic applications requiring precise quantification of FDP levels in diverse biological matrices
Catalog No. 50-000-50536 Supplier Cusabio Technology LLC Supplier No. CSBE07940H
This kit detects Fibrinogen Degradation Product (FDP) in human serum cell culture supernates urine and tissue homogenates using a quantitative sandwich ELISA method Designed for high sensitivity it offers a detection range of 6 25 ng/mL to 400 ng/mL with a lower limit of 1 56 ng/mL ensuring accurate measurement of low-abundance samples The assay requires a sample volume of 50-100 L and can be completed within 1-5 hours Absorbance is measured at 450 nm for result interpretation making it suitable for research and diagnostic applications requiring precise quantification of FDP levels in diverse biological matrices