Learn More
Diagnostic Biosystems FIBRINOGEN-FITC Fibrinogen-FITC
Supplier:  Diagnostic Biosystems F00601
Encompass_Preferred
This antibody reacts with native fibrinogen and with fibrinogen fragments D, E, X, and Y. Traces of contaminating antibodies have been removed by solid phase absorption with human plasma proteins. This antibody is well suited for tissue section staining but may also be used for other applications.
Catalog No. 50-240-7618