Learn More
Diagnostic Biosystems FIBRINOGEN-FITC Fibrinogen-FITC
Catalog No. 502407529
Encompass_Preferred
This antibody reacts with native fibrinogen and with fibrinogen fragments D, E, X, and Y. Traces of contaminating antibodies have been removed by solid phase absorption with human plasma proteins. This antibody is well suited for tissue section staining but may also be used for other applications.
Catalog No. 50-240-7529 Supplier Diagnostic Biosystems Supplier No. F006
This antibody reacts with native fibrinogen and with fibrinogen fragments D, E, X, and Y. Traces of contaminating antibodies have been removed by solid phase absorption with human plasma proteins. This antibody is well suited for tissue section staining but may also be used for other applications.