Learn More
Bioworld Fibrinogen from human plasma, 100 mg
Supplier:  Bioworld 220000011
Encompass_Preferred
Fibrinogen, also known as Factor I is a plasma protein that is essential for coagulation of blood and is coverted to fibrin by thrombinSpecifications:Synonym: Factor IForm: lyophilized powder (fibrous)50-70% protein λmax: 280nm
Catalog No. 50-252-961