Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fibrinogen gamma Chain Antibody (YA1896) | 10 UL
SDP

Catalog No. 5000444076
Encompass_Preferred

Fibrinogen gamma Chain Antibody (YA1896) is a rabbit-derived, non-conjugated IgG monoclonal antibody targeting the Fibrinogen gamma Chain. It is specifically developed for research use. This antibody is supplied as a liquid formulation, ensuring stability and convenience for various laboratory applications.

  • Host: Rabbit
  • Clonality: Recombinant, Monoclonal
  • Molecular weight: Predicted and observed band size 52 kDa
  • Species reactivity: Human
  • Applications: Western Blot (1:500-1:1000), Flow Cytometry (1:50-1:100)
  • Purity method: Affinity Chromatography
  • Conjugation: Non-conjugated
  • Isotype: IgG
  • Storage: Stored at -20°C for 1 year. Avoid repeated freeze/thaw cycles.

Catalog No. 50-004-44076 Supplier Medchemexpress LLC Supplier No. HYP8215110UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fibrinogen gamma Chain Antibody (YA1896) is a rabbit-derived, non-conjugated IgG monoclonal antibody targeting the Fibrinogen gamma Chain. It is specifically developed for research use. This antibody is supplied as a liquid formulation, ensuring stability and convenience for various laboratory applications.

  • Host: Rabbit
  • Clonality: Recombinant, Monoclonal
  • Molecular weight: Predicted and observed band size 52 kDa
  • Species reactivity: Human
  • Applications: Western Blot (1:500-1:1000), Flow Cytometry (1:50-1:100)
  • Purity method: Affinity Chromatography
  • Conjugation: Non-conjugated
  • Isotype: IgG
  • Storage: Stored at -20°C for 1 year. Avoid repeated freeze/thaw cycles.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.