Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fibrinogen gamma Chain Antibody (YA1896) | 100 UL
SDP

Catalog No. 5000441625
Encompass_Preferred

Fibrinogen gamma Chain Antibody (YA1896) is a Rabbit-derived, non-conjugated IgG monoclonal antibody that targets Fibrinogen gamma Chain. It is intended for research use only.

  • Host: Rabbit
  • Clonality: Recombinant, monoclonal
  • Species reactivity: Human
  • Immunogen: A synthesized peptide derived from human Fibrinogen gamma chain aa400-453/453
  • Applications: Western blot (WB) and Flow cytometry (FC)
  • Isotype: IgG
  • Formulation: Supplied in 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.01% sodium azide and 0.05% BSA
  • Storage and stability: Stored at -20°C for 1 year. Avoid repeated freeze/thaw cycles

Catalog No. 50-004-41625 Supplier Medchemexpress LLC Supplier No. HYP82151100UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fibrinogen gamma Chain Antibody (YA1896) is a Rabbit-derived, non-conjugated IgG monoclonal antibody that targets Fibrinogen gamma Chain. It is intended for research use only.

  • Host: Rabbit
  • Clonality: Recombinant, monoclonal
  • Species reactivity: Human
  • Immunogen: A synthesized peptide derived from human Fibrinogen gamma chain aa400-453/453
  • Applications: Western blot (WB) and Flow cytometry (FC)
  • Isotype: IgG
  • Formulation: Supplied in 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.01% sodium azide and 0.05% BSA
  • Storage and stability: Stored at -20°C for 1 year. Avoid repeated freeze/thaw cycles

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.