Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fibrinogen gamma chain protein | >95.0% | 20 UG
SDP

Catalog No. 5000294337
Encompass_Preferred

Recombinant human fibrinogen gamma chain (fragment V169-L453) produced in Pichia pastoris for research applications in coagulation, wound repair, and cell-matrix interaction studies. The protein is tag-free, glycosylated, and supplied as a filtered solution formulated with stabilizers for laboratory assays.

  • Recombinant human sequence (V169-L453).
  • Expressed in P. pastoris.
  • Tag-free and glycosylated.
  • Supplied in PBS pH 7.4 with 5% trehalose, 5% mannitol, and 0.01% Tween 80.
  • Purity >95% by reducing SDS-PAGE.
  • Approximate molecular weight 32.2 kDa.
  • Store at -80 °C; stable at -20 °C for 3 months after opening.

Catalog No. 50-002-94337 Supplier Medchemexpress LLC Supplier No. HYP7577220UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human fibrinogen gamma chain (fragment V169-L453) produced in Pichia pastoris for research applications in coagulation, wound repair, and cell-matrix interaction studies. The protein is tag-free, glycosylated, and supplied as a filtered solution formulated with stabilizers for laboratory assays.

  • Recombinant human sequence (V169-L453).
  • Expressed in P. pastoris.
  • Tag-free and glycosylated.
  • Supplied in PBS pH 7.4 with 5% trehalose, 5% mannitol, and 0.01% Tween 80.
  • Purity >95% by reducing SDS-PAGE.
  • Approximate molecular weight 32.2 kDa.
  • Store at -80 °C; stable at -20 °C for 3 months after opening.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.