Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fibrinogen gamma chain protein, human (P. pastoris) | >95.0% | 10 UG
SDP

Catalog No. 5000294409
Encompass_Preferred

Recombinant human fibrinogen gamma chain protein (V169-L453) expressed in P. pastoris. This tag-free protein is supplied as a filtered, stabilized solution for biochemical and cellular assays investigating fibrin formation, coagulation, and wound repair. Provided in microgram quantities for research use.

  • Recombinant human sequence V169-L453 expressed in P. pastoris.
  • Tag free, suitable for assays requiring unmodified protein.
  • Approximate molecular weight 32.2 kDa.
  • Purity >95.0% by reducing SDS-PAGE.
  • Supplied in PBS pH 7.4 with 5% trehalose, 5% mannitol, 0.01% Tween 80.
  • Stable when stored at -80°C; avoid repeated freeze-thaw cycles.
  • Available in small microgram sizes for research applications.

Catalog No. 50-002-94409 Supplier Medchemexpress LLC Supplier No. HYP7577210UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human fibrinogen gamma chain protein (V169-L453) expressed in P. pastoris. This tag-free protein is supplied as a filtered, stabilized solution for biochemical and cellular assays investigating fibrin formation, coagulation, and wound repair. Provided in microgram quantities for research use.

  • Recombinant human sequence V169-L453 expressed in P. pastoris.
  • Tag free, suitable for assays requiring unmodified protein.
  • Approximate molecular weight 32.2 kDa.
  • Purity >95.0% by reducing SDS-PAGE.
  • Supplied in PBS pH 7.4 with 5% trehalose, 5% mannitol, 0.01% Tween 80.
  • Stable when stored at -80°C; avoid repeated freeze-thaw cycles.
  • Available in small microgram sizes for research applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.