Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Fibrinogen

Catalog No. MP215416525 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215416525 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP215416525 Supplier MP Biomedicals™ Supplier No. 0215416525
Only null left
Add to Cart
Add to Cart

Converted to fibrin by thrombin. Fibrinogen, MP Biomedicals a is a blood protein that is involved in clotting. It consists of three non-identical pairs of disulfide bonded chains. Fibrinogen has been used in studies of haemostatic therapy in trauma patien

Fibrinogen has been used in studies of haemostatic therapy in surgical and massive trauma patients. These studies have shown that fibrinogen may prove to be more superior in stopping blood loss when compared to using fresh frozen plasma (FFP).

Fibrinogen is an acute phase protein that is part of the coagulation cascade of proteins. The end result of the cascade is the production of thrombin that converts fibrinogen to fibrin. Fibrinogen from any mammalian source will be cleaved by thrombin from any mammalian source.

  • Thrombin rapidly proteolyzes fibrinogen, releasing fibrinopeptide A
  • The loss of this small peptide is not sufficient to make the resulting fibrin molecule insoluble, but it tends to form complexes with adjacent fibrin and fibrinogen molecules
  • Thrombin then cleaves a second peptide, fibrinopeptide B, from fibrin and the fibrin monomers formed then polymerize spontaneously to form an insoluble gel
  • The polymerized fibrin is held together by noncovalent and electrostatic forces and stabilized by the transamidating enzyme, factor XIIIa, that is produced by the action of thrombin on factor XIII
  • The insoluble fibrin aggregates (clots) and aggregated platelets then block the damaged blood vessel and prevent further bleeding
  • The amount of fibrinogen in the plasma can serve as a nonspecific indicator of whether or not an inflammatory process is present in the body
TRUSTED_SUSTAINABILITY

Chemical Identifiers

Synonym Factor I

Specifications

Quantity 25 g
Content And Storage −20°C
Form Powder
Solubility Soluble in DI H2O (4% w/v).
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.