Learn More
MOLECULAR DEPOT LLC Fibrinogen Plasminogen -50 mg
Catalog No. 5000013567
Encompass_Preferred
Human Fibrinogen Solution Plasminogen Depleted consists of human fibrinogen depleted of plasminogen Fibrinogen (factor I) is a glycoprotein found in the blood of vertebrates It is directly involved in the complex process of blood clotting This fibrinogen has been depleted of plasminogen using affinity chromatography For in vitro research use only
Catalog No. 50-000-13567 Supplier MOLECULAR DEPOT LLC Supplier No. P2010023
Human Fibrinogen Solution Plasminogen Depleted consists of human fibrinogen depleted of plasminogen Fibrinogen (factor I) is a glycoprotein found in the blood of vertebrates It is directly involved in the complex process of blood clotting This fibrinogen has been depleted of plasminogen using affinity chromatography For in vitro research use only