Learn More
Alpco Ltd Porcine Fibrinogen ELISA
SDP
Supplier:  Alpco Ltd 41FIBPOE01
Encompass
The Porcine Fibrinogen ELISA is a highly sensitive two-site enzyme linked immunoassay (ELISA) for measuring Fibrinogen in biological samples of pigs. Soluble Fibrinogen circulates in the blood and provides the material from which the insoluble fibrin clot is formed during blood coagulation. Fibrinogen is an acute phase reactant that may be a useful marker for infection and inflammation. Research Use Only. Not for Use in Diagnostic Procedures.
Catalog No. 50-751-3576