Learn More
Innovative Research Inc Mouse Fibrinogen Purified FITC Labeled 1mg
Supplier:  Innovative Research Inc IMSFBGFITC1MG
Encompass_Preferred
Mouse Fibrinogen Purified FITC Labeled from Innovative Research has been purified from plasminogen depleted Mouse Plasma using a combination of several chromatographic and fractionation steps then labeled at primary amines with fluorescein isothiocyanate (FITC). This is a frozen liquid buffered in 0.02M Sodium Citrate-HCl; pH 7.4, with a concentration of 2.0 mg/ml and a purity of >95%.
Catalog No. 50-203-6541
 Restricted Item