Learn More
Pacific Hemostasis™ Fibrinogen Reference Plasma, 10 x 1mL
Screen and quantify plasma fibrinogen based on the Clauss method.
Catalog No. 22-176305
Item Discontinued This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.
This item is not returnable. View return policy
Description
Description
Screen and quantify plasma fibrinogen based on the Clauss method with Thermo Scientific™ Pacific Hemostasis™ Fibrinogen/Fibrin Analysis.
Fibrinogen Assay Set:
  • Quantifies plasma fibrinogen levels
  • Detects acquired and congenital hemorrhagic disorders
  • Components available separately
Thrombin Time Assay:
  • Provides qualitative determination
  • Useful in screening for fibrinogen disorders by measuring the formation of the fibrin clot
  • Reconstituted stability: 8 hours at 2° to 8°C
Ordering Information:
Products can be purchased as a set or individually.
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions