Learn More
ATCC Fibroblast Basal Medium
Catalog No. 502384719
Encompass
Fibroblast Basal Medium is a sterile, phenol-red-free, liquid tissue culture medium intended for use as one component in a complete ATCC Primary Cell Solutions system. This system is designed to support fibroblasts derived from various types of normal human tissue. Fibroblast Basal Medium contains essential and non-essential amino acids, vitamins, other organic compounds, trace minerals, and inorganic salts.
Catalog No. 50-238-4719 Supplier ATCC Supplier No. PCS201030
 Restricted Item
Add to Cart
Add to Cart
Fibroblast Basal Medium is a sterile, phenol-red-free, liquid tissue culture medium intended for use as one component in a complete ATCC Primary Cell Solutions system. This system is designed to support fibroblasts derived from various types of normal human tissue. Fibroblast Basal Medium contains essential and non-essential amino acids, vitamins, other organic compounds, trace minerals, and inorganic salts.