Learn More
Promocell GmbH PromoCell Fibroblast Basal Medium 2 500 ml
Catalog No. 50306224
Encompass
PromoCell Fibroblast Growth Medium 2 is a low-serum (2% V/V) medium and has been developed for the in vitro cultivation of adult Human Fibroblasts.Although all PromoCell media are optimized for use with primary human cells, we have received feedback from customers that this particular medium can also be used for bovine, porcine, and rat fibroblasts, as well as fibroblast cell
Catalog No. 50-306-224 Supplier Promocell GmbH Supplier No. C23220
PromoCell Fibroblast Growth Medium 2 is a low-serum (2% V/V) medium and has been developed for the in vitro cultivation of adult Human Fibroblasts.Although all PromoCell media are optimized for use with primary human cells, we have received feedback from customers that this particular medium can also be used for bovine, porcine, and rat fibroblasts, as well as fibroblast cell