Learn More
Promocell GmbH PromoCell Fibroblast Growth Medium 2 500 ml, ready-to-use, with C-39325 attached to the flask
Catalog No. 50306213
Encompass
PromoCell Fibroblast Growth Medium 2 is a low-serum (2% V/V) medium and has been developed for the in vitro cultivation of adult Human Fibroblasts.Although all PromoCell media are optimized for use with primary human cells, we have received feedback from customers that this particular medium can also be used for bovine, porcine, and rat fibroblasts, as well as fibroblast cell lines.| Media supplement | Final supplement concentration||----------------------------------------------------|---------------------------------------------------------------|| Fetal Calf Serum | 0.02 ml / ml || Basic Fibroblast Growth Factor (recombinant human) | 1 ng / ml || Insulin (recombinant human) | 5 μg / ml |
Catalog No. 50-306-213 Supplier Promocell GmbH Supplier No. C23020
Add to Cart
Add to Cart
PromoCell Fibroblast Growth Medium 2 is a low-serum (2% V/V) medium and has been developed for the in vitro cultivation of adult Human Fibroblasts.Although all PromoCell media are optimized for use with primary human cells, we have received feedback from customers that this particular medium can also be used for bovine, porcine, and rat fibroblasts, as well as fibroblast cell lines.| Media supplement | Final supplement concentration||----------------------------------------------------|---------------------------------------------------------------|| Fetal Calf Serum | 0.02 ml / ml || Basic Fibroblast Growth Factor (recombinant human) | 1 ng / ml || Insulin (recombinant human) | 5 μg / ml |