Learn More
Promocell GmbH PromoCell Fibroblast Growth Medium 500 ml, ready-to-use, serum-free, with C-39315 attached to the flask
Catalog No. 50306212
Encompass
PromoCell Fibroblast Growth Medium is optimized for the cultivation of neonatal and juvenile fibroblasts.Although all PromoCell media are optimized for use with primary human cells, we have received feedback from customers that this particular medium can also be used for bovine, porcine, and rat fibroblasts as well as fibroblast cell lines.While the supplements of the Fibroblast Growth Medium are serum-free, the Basal Medium is manufactured with 0.5% serum.| Media supplement | Final supplement concentration||----------------------------------------------------|---------------------------------------------------------------|| Basic Fibroblast Growth Factor (recombinant human) | 1 ng / ml || Insulin (recombinant human) | 5 μg / ml |
Catalog No. 50-306-212 Supplier Promocell GmbH Supplier No. C23010
PromoCell Fibroblast Growth Medium is optimized for the cultivation of neonatal and juvenile fibroblasts.Although all PromoCell media are optimized for use with primary human cells, we have received feedback from customers that this particular medium can also be used for bovine, porcine, and rat fibroblasts as well as fibroblast cell lines.While the supplements of the Fibroblast Growth Medium are serum-free, the Basal Medium is manufactured with 0.5% serum.| Media supplement | Final supplement concentration||----------------------------------------------------|---------------------------------------------------------------|| Basic Fibroblast Growth Factor (recombinant human) | 1 ng / ml || Insulin (recombinant human) | 5 μg / ml |