Learn More
Promocell GmbH PromoCell Fibroblast Growth Medium 3 KIT 500 ml, Contains: C-23230, C-39350
Catalog No. 50306219
Encompass
PromoCell Fibroblast Growth Medium 3 has been developed for the in vitro cultivation of adult Human Cardiac Fibroblasts (HCF).Although all PromoCell media are optimized for use with primary human cells, we have received feedback from customers that this particular medium can also be used for porcine cardiac fibroblasts.| Media supplement | Final supplement concentration|| Fetal Calf Serum | 0.1 ml / ml || Basic Fibroblast Growth Factor (recombinant human) | 1 ng / ml || Insulin (recombinant human) | 5 μg / ml |
Catalog No. 50-306-219 Supplier Promocell GmbH Supplier No. C23130
PromoCell Fibroblast Growth Medium 3 has been developed for the in vitro cultivation of adult Human Cardiac Fibroblasts (HCF).Although all PromoCell media are optimized for use with primary human cells, we have received feedback from customers that this particular medium can also be used for porcine cardiac fibroblasts.| Media supplement | Final supplement concentration|| Fetal Calf Serum | 0.1 ml / ml || Basic Fibroblast Growth Factor (recombinant human) | 1 ng / ml || Insulin (recombinant human) | 5 μg / ml |