Learn More
Sigma Aldrich Fine Chemicals Biosciences fibroblast Growth factor 1
Catalog No. 502904096
Encompass
fGf10 (fibroblast growth factor 10) is a member of the fGf protein family which is a mitogen of epithelial origin and has a paracrine activity. It is produced by mesenchymal lineage cells and acts as a ligand for fGfR most notably fGfR2b which is expressed by epithelial cells.
Catalog No. 50-290-4096 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. F892425UG
fGf10 (fibroblast growth factor 10) is a member of the fGf protein family which is a mitogen of epithelial origin and has a paracrine activity. It is produced by mesenchymal lineage cells and acts as a ligand for fGfR most notably fGfR2b which is expressed by epithelial cells.