Learn More
Cusabio Technology LLC Fibroblast growth factor 15
Catalog No. 5000051451
Encompass_Preferred
The Mouse Fibroblast Growth Factor 15 (FGF15) ELISA Kit is designed for the quantitative detection of FGF15 in serum plasma tissue homogenates and cell lysates This sandwich-format assay offers a broad detection range of 15 6 pg/ml to 1000 pg/ml with a sensitivity of 3 9 pg/ml ensuring precise measurement of low-abundance samples The protocol requires a short incubation time of 1-5 hours and a minimal sample volume of 50-100 l per test Results are determined via absorbance readings at 450 nm providing reliable and reproducible data for research applications The kit is optimized for efficiency and accuracy making it suitable for studies involving FGF15 regulation signaling pathways and related physiological or pathological processes
Catalog No. 50-000-51451 Supplier Cusabio Technology LLC Supplier No. CSBEL522052MO
The Mouse Fibroblast Growth Factor 15 (FGF15) ELISA Kit is designed for the quantitative detection of FGF15 in serum plasma tissue homogenates and cell lysates This sandwich-format assay offers a broad detection range of 15 6 pg/ml to 1000 pg/ml with a sensitivity of 3 9 pg/ml ensuring precise measurement of low-abundance samples The protocol requires a short incubation time of 1-5 hours and a minimal sample volume of 50-100 l per test Results are determined via absorbance readings at 450 nm providing reliable and reproducible data for research applications The kit is optimized for efficiency and accuracy making it suitable for studies involving FGF15 regulation signaling pathways and related physiological or pathological processes