Learn More
Creative Biomart Fibroblast Growth Factor 2 (rFGF 2)
Catalog No. 502084322
Encompass_Preferred
bFGF Rat Recombinant (FGF-2) produced in E Coli is a single, non-glycosylated, polypeptide chain containing 145 amino acids and having a molecular mass of 16.3 kDa.The bFGF2 is purified by proprietary chromatographic techniques.
Catalog No. 50-208-4322 Supplier Creative Biomart Supplier No. THP0271
bFGF Rat Recombinant (FGF-2) produced in E Coli is a single, non-glycosylated, polypeptide chain containing 145 amino acids and having a molecular mass of 16.3 kDa.The bFGF2 is purified by proprietary chromatographic techniques.