Learn More
Creative Biomart Velafermin, FGF20
Catalog No. 502084253
Encompass_Preferred
The product was known as fibroblast growth factor-20 (FGF-20), which promotes both epithelial and mesenchymal cell proliferation in vitro and has demonstrated activity in multiple animal models.
Catalog No. 50-208-4253 Supplier Creative Biomart Supplier No. THP0202
The product was known as fibroblast growth factor-20 (FGF-20), which promotes both epithelial and mesenchymal cell proliferation in vitro and has demonstrated activity in multiple animal models.