Learn More
Cusabio Technology LLC fibroblast growth factor 23
Supplier:  Cusabio Technology LLC CSBEL008629MO
Encompass_Preferred
This kit detects FGF23 in mouse serum plasma and tissue homogenates using a quantitative sandwich ELISA method offering a detection range of 23 44 pg/mL to 1500 pg/mL with a sensitivity of 5 86 pg/mL The assay requires 50-100 L of sample per test and can be completed within 1-5 hours utilizing absorbance measurement at 450 nm for precise quantification Designed for reliable performance across biological matrices it provides researchers with a robust tool to measure FGF23 concentrations in experimental models while maintaining analytical accuracy throughout the working range
Catalog No. 50-000-51201