Learn More
Sigma Aldrich Fine Chemicals Biosciences fibroblast Growth factor 4
Catalog No. 502904664
Encompass
fibroblast Growth factor-4 (fGf4) is a 19 kDa protein belonging to the fGf family of proteins. fibroblast growth factors (fGf) are glycoproteins concealed in the extracellular matrix and cell surface. There are around 22 different types of fGfs known. These factors weigh in the range of 17 to 34 kDa and they possess a conserved sequence of 120 amino acids. The fGf4 gene is mapped to human chromosome 11q13.3.
Catalog No. 50-290-4664 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. F842425UG
fibroblast Growth factor-4 (fGf4) is a 19 kDa protein belonging to the fGf family of proteins. fibroblast growth factors (fGf) are glycoproteins concealed in the extracellular matrix and cell surface. There are around 22 different types of fGfs known. These factors weigh in the range of 17 to 34 kDa and they possess a conserved sequence of 120 amino acids. The fGf4 gene is mapped to human chromosome 11q13.3.